Would you like to log in again?
YPG Agar, HiMedia
Supplier: HiMedia Laboratories
M1368-500G M1368-500G M1368-5KG M1368-5KG M1368-2.5KG M1368-2.5KG
89223-876EA 842.39 USD InStock
89223-876 89223-878 CA89223-876 CA89223-878 95021-028 CA95021-028
YPG Agar, HiMedia
Media for Microbiology Dehydrated Media for Microbiology
YPG Agar is recommended for the growth of Saccharomyces cerevisiae for molecular biology purpose.
General methods in yeasts genetics specify using yeast extract, peptone medium for cultivating Saccharomyces cerevisiae
and other yeasts. The yeast extract allows faster growth so that during exponential or log-phase growth, the cells divide every
90 minutes. Peptic digest of animal tissue is as a source of carbon, nitrogen, vitamins and minerals.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty