Would you like to log in again?
Christensen Citrate Agar, HiMedia Laboratories
Supplier: HiMedia Laboratories
M143-500G M143-2.5KG M143-5KG M143-500G M143-2.5KG M143-5KG
89224-030EA 1246.29 USD InStock
CA89224-030 89224-030 CA61000-026 CA89224-028 61000-026 89224-028
Christensen Citrate Agar, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
For the differentiation of enteric pathogens and coliforms on the basis of citrate utilization.
  • Light yellow to light pink homogeneous free flowing powder
  • Reaction of 2.48% w/v aqueous solution at 25 °C. pH : 6.9 ± 0.2
Store below 30 °C in tightly closed container and the prepared medium at 2 - 8 °C.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty