Would you like to log in again?
Rapid HiColiform Broth Dehydrated Culture Media, HiMedia Laboratories
Supplier: HiMedia Laboratories
M1453-100G M1453-2.5KG M1453-500G M1453-5KG M1453-2.5KG M1453-500G M1453-5KG
89224-066EA 2063.73 USD InStock
CA89224-064 89224-066 89224-064 CA95021-150 75840-336 75840-332 CA89224-066
Rapid HiColiform Broth Dehydrated Culture Media, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
Rapid Hicoliform is used for the detection and confirmation of Escherichia coli and total coliforms on the basis of enzyme substrate reaction from water samples using a combination of chromogenic and fluorogenic substrate.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty