Would you like to log in again?
Acetamide Broth (Twin Pack), HiMedia Laboratories
Supplier: HiMedia Laboratories
M148-500G M148-500G M148-2.5KG M148-2.5KG M148-5KG M148-5KG
61000-030EA 410.01 USD InStock
CA61000-030 CA89224-130 CA89224-140 61000-030 89224-130 89224-140
Acetamide Broth (Twin Pack), HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
Acetamide broth is used for the confirmation of Pseudomonas aeruginosa in water samples.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty