Would you like to log in again?
Rappaport Vassiliadis Soya Broth (RVS Broth), HiMedia Laboratories
Supplier: HiMedia Laboratories
M1491-500G M1491-5KG M1491-5KG M1491-500G M1491-2.5KG
89224-160EA 1518.02 USD InStock
CA89224-160 89224-160 95021-224 CA95021-224 89224-158
Rappaport Vassiliadis Soya Broth (RVS Broth), HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
Rappaport Vassiliadis Soya Broth (RVS Broth) is recommended as a selective enrichment medium for the Salmonellae species from the food and animal feeding stuffs.
RVS broth medium can also be used in direct enrichment of samples containing low inoculum.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty