Would you like to log in again?
Buffered Peptone Water, Dehydrated Culture Media, HiMedia Laboratories
Supplier: HiMedia Laboratories
M1494I-500G M1494I-25KG M1494I-10KG M1494I-5KG M1494I-5KG M1494I-2.5KG M1494I-2.5KG M1494I-500G M1494I-10KG M1494I-25KG
89224-166EA 2758.85 USD InStock
89224-166 CA89224-164 CA89224-162 89224-164 89224-162 CA89224-168 95021-226 CA95021-226 89224-168 CA89224-166
Buffered Peptone Water, Dehydrated Culture Media, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
A pre-enrichment medium for increasing the recovery of injured Salmonella species from foods prior to selective enrichment and isolation.
Certifications: ISO 6579-2017 , ISO 6887 and ISO 21528-2017.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty