Would you like to log in again?
Arabinose Agar Base, HiMedia Laboratories
Supplier: HiMedia Laboratories
M1576-2.5KG M1576-5KG M1576-2.5KG M1576-5KG
89224-286EA 1587.6 USD InStock
CA89224-284 89224-286 89224-284 CA89224-286
Arabinose Agar Base, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
Recommended to differentiate enterococcus faecium from Enterococcus faecalis.
  • Multiple sizes available
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty