Would you like to log in again?
Bolton Broth Base, Dehydrated Culture Media, HiMedia Laboratories
Supplier: HiMedia Laboratories
M1592-5KG M1592-2.5KG M1592-2.5KG M1592-5KG M1592-500G M1592-500G
89224-340EA 1808.93 USD InStock
CA89224-340 CA95021-316 89224-340 CA89224-338 89224-338 95021-316
Bolton Broth Base, Dehydrated Culture Media, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
For the selective enrichment of Campylobacter species from food and animal foodstuffs.
  • Microbiological culture media
  • pH: 7.20 - 7.60
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty