Would you like to log in again?
Skim Milk PCA, HiMedia
Supplier: HiMedia Laboratories
M1623-5KG M1623-500G M1623-2.5KG M1623-2.5KG M1623-500G M1623-5KG
89224-450EA 2047.38 USD InStock
CA89224-450 89224-450 CA89224-448 89224-448 95021-368 CA95021-368
Skim Milk PCA, HiMedia
Media for Microbiology Dehydrated Media for Microbiology
Skim milk plate count agar is recommended for determining the microbial count in milk and dairy products.
  • Appearance: Cream to yellow homogeneous free flowing powder
  • Gelling: Firm, comparable with 1.05% agar gel
Casein enzymic hydrolysate provides amino acids and other complex nitrogenous substances. Yeast extract supplies vitamin B complex. Addition of skim milk in the medium makes the conditions optimal for microorganisms which grow in milk. A wide range of microorganisms can be cultured and enumerated on this medium.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty