Would you like to log in again?
Phenol Red Sucrose Broth, HiMedia Laboratories
Supplier: HiMedia Laboratories
M274-2.5KG M274-500G M274-5KG M274-2.5KG M274-500G M274-5KG
95021-592EA 130.96 USD InStock
95021-592 CA95021-592 CA89224-998 CA89224-996 89224-998 89224-996
Phenol Red Sucrose Broth, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
Phenol Red Sucrose Broth is used for sucrose fermentation studies of microorganisms.
Phenol Red Broth Medium is formulated and is recommended to determine the fermentation reaction of carbohydrates for the differentiation of micro organisms
Proteose peptone and beef extract serve as sources for carbon and nitrogen. Sodium chloride is the osmotic stabilizer. Phenol red is the pH indicator, which turns yellow at acidic pH i.e. on fermentation of sucrose. Gas formation is seen in Durhams tubes. All of the Enterobacteriaceae grow well in this medium. In addition to producing a pH colour shift, the production of mixed acids, notably butyric acids, often results in a pungent, foul odour from the culture medium.
This medium with various carbohydrates serves as a differential medium by aiding in differentiation of various species and genera by their ability to ferment the specific carbohydrate, with the production of acid or acid and gas.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty