Would you like to log in again?
Wort Broth, HiMedia Laboratories
Supplier: HiMedia Laboratories
M333-500G M333-5KG M333-2.5KG
95021-694EA 193.69 USD InStock
95021-694 89225-200 89225-202
Wort Broth, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
For the cultivation and enumeration of yeasts.
  • Phosphate buffer
  • Propagate the multiplication of yeasts
  • High acidic pH inhibits many bacteria
Wort Broth is formulated for the cultivation of fungi especially yeasts in syrups and butter. Wort Broth is especially designed to propagate the multiplication of yeasts and often it has been employed as a semi-selective or enrichment medium. Yeast grows well in culture media containing dextrin or maltose in acidic environment. In this medium peptic digest of animal tissue and malt extract provide nitrogenous and other nutrients for the growth of yeasts. Dextrin and maltose are fermentable carbohydrates. The high acidic pH inhibits many bacteria. Phosphate buffers the medium.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty