Would you like to log in again?
Bushnell Haas Agar, HiMedia Laboratories
Supplier: HiMedia Laboratories
M349-5KG M349-2.5KG M349-2.5KG M349-500G M349-500G M349-5KG
89225-252EA 975.71 USD InStock
89225-252 CA61000-080 61000-080 CA89225-252 89225-254 CA89225-254
Bushnell Haas Agar, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
Bushnell Haas Agar are recommended for the microbial examination of fuels and for studying microbial hydrocarbon deterioration.
  • White to cream homogeneous free flowing powder
  • Firm gelling, comparable with 2.0% agar gel
  • pH: 6.80 to 7.20
  • Store below 30 °C
Bushnell Haas Agar contains magnesium sulfate (0.200 g/l); calcium chloride (0.020 g/l); monopotassium phosphate (1.000 g/l); dipotassium phosphate (1.000 g/l); ammonium nitrate (1.000 g/l); ferric chloride (0.050 g/l) and agar (20.000 g/l).
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty