Would you like to log in again?
ISP Medium No. 5, Glycerol Asparagine Agar Base, HiMedia Laboratories
Supplier: HiMedia
M360-2.5KG M360-5KG M360-2.5KG M360-5KG M360-500G M360-500G
89225-294EA 2463.71 USD InStock
CA89225-292 89225-294 CA89225-294 89225-292 CA61000-088 61000-088
ISP Medium No. 5, Glycerol Asparagine Agar Base, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
Used for cultivation of Streptomyces species as per International Streptomyces project.
  • Provides consistent and reproducible characteristic features of Streptomyces
  • Dipotassium phosphate buffers the medium
  • Based on the formulation described by Shirling and Gottlieb
ISP Medium No
This medium provides consistent and reproducible characteristic features of Streptomyces. Glycerol serves as the carbon source while asparagine is the amino acid source for the growth of Streptomyces species. Trace mineral requirement of Streptomyces is satisfied by the trace salt solution, which contains various salts. Dipotassium phosphate buffers the medium.
5 (Glycerol Asparagine Agar Base) is based on the formulation described by Shirling and Gottlieb and is used for cultivation and characterization of Streptomyces species as recommended by the International Streptomyces Project. Being primarily soil inhabitants, Streptomyces are most commonly limited to causing actinomycotic mycetoma. Areas more prone to formation of mycetomas are those that are frequently traumatized or that come into contact with soil.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty