Would you like to log in again?
Zobell Marine Agar and Broth, HiMedia Laboratories
Supplier: HiMedia Laboratories
M385-10KG M385-500G M384-2.5KG M385-2.5KG M385-10KG M384-5KG M385-500G M384-500G M385-5KG M384-2.5KG M385-5KG M384-5KG M385-2.5KG M384-500G
89225-372EA 655.05 USD InStock
89225-372 CA97065-952 CA89225-372 89225-374 97065-952 CA97065-950 97065-950 95021-752 CA89225-374 95021-754 89225-376 CA95021-752 CA95021-754 CA89225-376
Zobell Marine Agar and Broth, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
Zobell Marine Agar is used in the cultivation of heterotrophic marine bacteria, while Zobell Marine Broth is used in the isolation and enumeration of heterotrophic marine bacteria.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty