Would you like to log in again?
Mueller Hinton Broth, HiMedia Laboratories
Supplier: HiMedia Laboratories
M391-5KG M391-2.5KG M391-5KG M391-2.5KG
89225-394EA 703.02 USD InStock
89225-394 89225-396 CA89225-394 CA89225-396
Mueller Hinton Broth, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
Used to determine the susceptibility of bacteria to Sulphonamides by the tube dilution method.
  • Multiple sizes available
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty