Would you like to log in again?
GC Agar Base Dehydrated Culture Media, HiMedia
Supplier: HiMedia Laboratories
M434-500G M434-2.5KG
89225-518EA 630.64 USD InStock
CA95021-824 89225-518
GC Agar Base Dehydrated Culture Media, HiMedia
Media for Microbiology Dehydrated Media for Microbiology
For the selective isolation and cultivation of gonococci.
  • Microbiological culture media
  • Supplied in poly bottles
Requires added blood or hemoglobin and other supplements.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty