Would you like to log in again?
HiVeg™ Wagatsuma Agar Base, HiMedia Laboratories
Supplier: HiMedia
HiVeg™
M626-500G M626-5KG M626-5KG M626-2.5KG M626-2.5KG M626-500G
89226-060EA 803.64 USD InStock
89226-060 CA95022-100 CA89226-062 95022-100 89226-062 CA89226-060
HiVeg™ Wagatsuma Agar Base, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
Wagatsum agar base is recommended for the performance of Kanagawa test to identify virulent vibrio parahaemolyticus strains.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty