Would you like to log in again?
Yeast Mannitol Agar with Congo Red, HiMedia Laboratories
Supplier: HiMedia Laboratories
M721-5KG M721-2.5KG M721-5KG M721-500G M721-500G M721-2.5KG
61000-170EA 220.76 USD InStock
CA61000-170 61000-170 89226-244 CA89226-246 CA89226-244 89226-246
Yeast Mannitol Agar with Congo Red, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
Yeast Mannitol Agar is used for the cultivation of soil microorganisms like Rhizobium species.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty