Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Wilkins Chalgren Anaerobic Broth Base, HiMedia Laboratories

Supplier: HiMedia Laboratories
M863-500G M863-2.5KG M863-2.5KG M863-5KG M863-500G M863-5KG
89226-616EA 780.54 USD InStock
89226-616 CA95022-378 CA89226-618 95022-378 CA89226-616 89226-618
Wilkins Chalgren Anaerobic Broth Base, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
Wilkins chalgren anaerobic broth base is used for cultivation and susceptibility testing of anaerobic bacteria.

  • pH: 6.90 to 7.30
  • Appearance: Cream to yellow homogeneous free flowing powder

The broth medium is also usefull for broth micro-dilution tests.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty