Would you like to log in again?
PSB Broth Base, HiMedia Laboratories
Supplier: HiMedia Laboratories
M941-5KG M941-2.5KG M941-5KG M941-2.5KG M941-500G M941-500G
95022-502EA 203.81 USD InStock
95022-502 CA89226-888 CA89226-886 89226-888 89226-886 CA95022-502
PSB Broth Base, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
PSB broth base is used for primary enrichment and enumeration of Yersinia enterocolitica from foods.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty