Would you like to log in again?
Tryptone Peptone Glucose Yeast Extract Broth Base, HiMedia Laboratories
Supplier: HiMedia Laboratories
M969-500G M969-5KG M969-500G M969-2.5KG M969-2.5KG M969-5KG
95022-554EA 220.73 USD InStock
CA89227-006 CA95022-554 95022-554 89227-004 89227-006 CA89227-004
Tryptone Peptone Glucose Yeast Extract Broth Base, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
This broth base without trypsin is used to test toxicity type of Clostridium botulinum cultures.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty