Would you like to log in again?
XLD HiVeg™ Agar, Himedia
Supplier: HiMedia Laboratories
HiVeg™
MV031-5KG MV031-500G MV031-2.5KG MV031-5KG MV031-500G MV031-2.5KG
95044-414EA 182.15 USD InStock
CA89227-258 CA95044-414 95044-414 89227-256 89227-258 CA89227-256
XLD HiVeg™ Agar, Himedia
Media for Microbiology Dehydrated Media for Microbiology
Recommended for the selective isolation and enumeration of Salmonella serotype Typhi and other Salmonella species.
  • Can be used as a diagnostic aid in the identification of Enterobacteriaceae
Ideal medium for screening samples containing mixed flora of enteric pathogens as recovery of Salmonellae and Shigellae is not conspicuous by even profuse growth of other species (2, 3).
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty