Would you like to log in again?
Deoxycholate Citrate Agar, HiVeg™, HiMedia Laboratories
Supplier: HiMedia Laboratories
MV065-500G MV065-5KG MV065-2.5KG MV065-500G MV065-2.5KG MV065-5KG
61000-328EA 185.56 USD InStock
CA89227-346 CA61000-328 61000-328 89227-344 CA89227-344 89227-346
Deoxycholate Citrate Agar, HiVeg™, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
Deoxycholate citrate agar is a selective medium recommended for the isolation of enteric pathogens particularly Salmonella and Shigella species.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty