Would you like to log in again?
Brucella HiVeg™ Agar Base Dehydrated Culture Media, HiMedia Laboratories
Supplier: HiMedia Laboratories
MV074-2.5KG MV074-500G MV074-500G MV074-5KG MV074-2.5KG MV074-5KG
89227-372EA 757.97 USD InStock
89227-372 CA89227-372 89227-374 CA89227-374 CA61000-342 61000-342
Brucella HiVeg™ Agar Base Dehydrated Culture Media, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
Brucella HiVeg agar base with supplement is recommended for enrichment, isolation and cultivation of Brucella or Campylobacter species from clinical and nonclinical specimens.
Brucella HiVeg agar base is prepared by completely replacing animal based peptones with vegetable peptones, making the medium BSE/TSE risks free. Brucella are intracellular parasites that cause epizootic abortions in animals and septicemic febrile illness or localized infections of bone, tissue or organ systems in humans. Brucella species are highly fastidious and therefore require a nutrient rich medium to be able to grow. Also, Brucella species are highly infective and so extreme care should be taken while handling.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty