Would you like to log in again?
HiVeg™ Lysine Decarboxylase Broth, HiMedia Laboratories
Supplier: HiMedia Laboratories
HiVeg™
MV376-2.5KG
89228-566EA 528.3 USD InStock
89228-566
HiVeg™ Lysine Decarboxylase Broth, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
For differentiating Salmonella Arizonae from the Bethesda Ballerup group of Enterobacteriaceae.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty