Would you like to log in again?
HiVeg™ Motility Indole Ornithine Medium, HiMedia Laboratories
Supplier: HiMedia Laboratories
MV378-500G MV378-500G MV378-2.5KG MV378-5KG MV378-2.5KG MV378-5KG
89228-576EA 1557.85 USD InStock
CA89228-576 89228-576 89228-574 CA61000-944 61000-944 CA89228-574
HiVeg™ Motility Indole Ornithine Medium, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
Motility Indole Ornithine Medium is used for the identification of Enterobacteriaceae on the basis of motility, indole production and ornithine decarboxylase activity.
  • Light yellow coloured, may have slightly greenish tinge, homogeneous, free flowing powder
  • Storage below 30 °C
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty