Would you like to log in again?
Violet Red Glucose Agar, HiVeg™, HiMedia Laboratories
Supplier: HiMedia Laboratories
MV581-500G MV581-5KG MV581-2.5KG MV581-5KG MV581-500G MV581-2.5KG MV581A-500G MV581A-500G
61001-150EA 182.7 USD InStock
61001-150 CA61001-150 61001-152 CA89228-910 CA89228-912 89228-912 89228-910 CA61001-152
Violet Red Glucose Agar, HiVeg™, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
Violet Red Glucose HiVeg™ Agar is recommended for enumeration of Enterobacteriaceae in raw foods.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty