Would you like to log in again?
HiVeg™ Broth Base, HiMedia Laboratories
Supplier: HiMedia Laboratories
HiVeg™
MV936-5KG MV936-500G MV936-5KG MV936-500G MV936-2.5KG MV936-2.5KG
61001-472EA 247.93 USD InStock
CA89229-436 61001-472 CA89229-434 89229-434 CA61001-472 89229-436
HiVeg™ Broth Base, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
Potassium cyanide HiVeg™ Broth Base with KCN supplementation is used for differentiation of the members of Enterobacteriaceae on the basis of potassium cyanide tolerance.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty