Would you like to log in again?
HiVeg™ Streptococcus thermophilus Isolation Agar, HiMedia Laboratories
Supplier: HiMedia
HiVeg™
MV948-5KG MV948-500G MV948-500G MV948-2.5KG MV948-5KG MV948-2.5KG
61001-482EA 294.42 USD InStock
61001-482 89229-460 CA89229-460 CA89229-458 CA61001-482 89229-458
HiVeg™ Streptococcus thermophilus Isolation Agar, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
Used for determining the ratio of Streptococcus thermophilus and Lactobacillus bulgaricus in yoghurt.
  • Reconstitution: 42.0 g/l
  • Sterilization: 121 °C/15 minutes
Caution: Storage conditions: dry medium - below 30 °C, prepared medium 2 to 8 °C.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty