Would you like to log in again?
D(+) Maltose, Monohydrate, HiMedia Laboratories
Supplier: HiMedia Laboratories
RM018-500G RM018-2.5KG RM018-500G RM018-10KG RM018-25KG RM018-5KG
89229-642EA 2678.92 USD InStock
89229-642 89229-648 61001-554 CA61001-554 89229-644 89229-646
D(+) Maltose, Monohydrate, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
Maltose is a sugar. It contains one molecule of water of hydration or is anhydrous used for bacteriological purpose.
  • Apperance: White crystalline powder with a slightly sweet taste
  • Solubility: Freely soluble in boiling distilled water, slightly soluble in alcohol and insoluble in chloroform and ether
  • Clarity: 5% w/v solution in boiling distilled water is clear and colorless
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty