Would you like to log in again?
Yeast Extract (Paste), HiMedia Laboratories
Supplier: HiMedia Laboratories
RM025-2.5KG RM025-10KG RM025-25KG RM025-5KG
89229-706EA 274.92 USD InStock
89229-706 89229-708 89229-704 89229-710
Yeast Extract (Paste), HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
Yeast extract paste suitable for cultivation of various microorganisms, fermentation and other biological products.
  • Brownish yellow to brown homogenous highly viscous paste, having characteristic odor but not putrescent
  • Freely soluble in distilled/purified water, insoluble in alcohol
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty