Would you like to log in again?
Casein Peptone, HiMedia
Supplier: HiMedia Laboratories
RM714-25KG RM714-500G RM714-5KG RM714-10KG RM714-500G RM714-10KG RM714-2.5KG RM714-5KG RM714-2.5KG RM714-25KG
89229-970EA 693.06 USD InStock
89229-970 CA89229-970 89229-972 CA89229-968 61001-642 CA61001-642 89229-968 89229-974 CA89229-974 CA89229-972
Casein Peptone, HiMedia
Media for Microbiology Dehydrated Media for Microbiology
Casein peptone is used to support the growth of fastidious microorganisms and is suitable for use in industrial fermentation.
  • Enzymic digest of milk protein
  • Nitrogen source for microorganisms
  • Suitable for use in industrial fermentation
HiCase Peptone is enzymic digest of milk protein. It is manufactured under controlled conditions. Milk protein being a rich source of amino nitrogen, the hydrolysed product HiCase Peptone obtained also contains peptides, aminoacids etc. which acts as nitrogen source for microorganisms when used in media formulations.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty