Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ammonium iron (III) citrate, Reagent Grade

Supplier: VWR
Synonyms: Ammonium iron (III) citrateFerric Ammonium CitrateAmmonium ferric citrate

VWRV0846-100G VWRV0846-500G
89230-024EA 79.85 USD InStock
89230-024 89230-026
Ammonium iron (III) citrate, Reagent Grade
Ammonium iron (III) citrate


Formula: C₆H₅+4y)Fe(x)N(y)O₇
Storage Temperature: Ambient
MDL Number: MFCD00013099
CAS Number: 1185-57-5
EINECS: 214-686-6
Order Now
Specification Test Results
Arsenic = 0.0004%
Iron (III) 20.5% - 22.5%
Lead = 0.003%
Solubility (5.0%, Water) Pass
Zinc = 0.1%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty