Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium gluconate 98-102%, crystals, high purity

Supplier: VWR
Synonyms: Sodium D-gluconateD-Gluconic acid sodium saltGluconic acid, sodium saltSodium (2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoate

VWRVE754-500G
89230-072CS 185.68 USD InStock
89230-072
Sodium gluconate 98-102%, crystals, high purity
Sodium gluconate


Formula: HOH₂C(CH(OH))₄COONa
MW: 218.14 g/mol
Melting Pt: 170…175 °C
Storage Temperature: Ambient
MDL Number: MFCD00064210
CAS Number: 527-07-1
EINECS: 208-407-7
Merck Index: 12,08766
https://us-legacy.vwr.com/stibo/web/std.lang.all/90/06/2299006.jpg
Order Now
Specification Test Results
Assay 98 % - 102 %
Heavy Metals 0.001 %
Lead 0.001 %
pH (10%, Water) 6.8 - 7.5

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty