Would you like to log in again?
Barnstead™ GenPure™ Water Purification Systems, Thermo Scientific
Supplier: Thermo Scientific
50131217
89230-358EA 17284.9 USD InStock
89230-358
Barnstead™ GenPure™ Water Purification Systems, Thermo Scientific
Water Purification Systems Pure and Ultra Pure Water Systems
Suitable for even the most demanding and sensitive applications, the family of Thermo Scientific™ Barnstead™ GenPure™ water purification systems deliver ultrapure 18,2 MΩ.cm water with consistent quality.
  • UV oxidation reduces organic levels to <5 ppb, and protects against microorganisms (only on systems containing a UV lamp (UV))
  • Ultrafiltration removes pyrogens from the feed water
  • Feed water monitoring alerts users to fluctuations in feed water quality
  • Real-time monitor of the amount of organic materials in the product water shown on the display
  • USP-compliant conductivity measurement with temperature compensation can be switched on or off
  • Feed water source: Potable tap water, pretreated by reverse osmosis, ion exchange or distillation
  • RS-232 interface enables data printing and satisfies GLP guidelines
  • User-friendly, illuminated four-line alpha-numeric displays and tilting control panels for optimal view
Special feature of GenPure Pro system:
- Volumetric dispensing: Dispense at the push of a button; control from 0,01-65 liters with an accurancy of <0,5%
Special features of GenPure xCAD Plus system:
- Volumetric dispensing: Dispense at the push of a button; control from 0,01-65 liters with an accurancy of <0,5%
- GenPure xCAD Plus remote dispenser for full control of the system
- Under-the-bench mounting: Ideal solution when bench space is limited
Ideal for molecular biology, microbiology and analytical chemistry applications (e.g. cell and tissue culture, PCR, DNA sequencing, electrophoresis, analytical chemistry, HPLC, CG, CG-MS, ICP-MS, AA, TOC Measurements).
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
SERVICES
Services: Thermo Scientific provides several service options for Thermo Barnstead Water System such as Installation, Extended Warranty, Extended Warranty plus on-site Annual Preventative Maintenance, and Total Care. To learn more about these services please search "Thermo Barnstead Water System Technical Services" in the VWR search field.
In addition, VWRCATALYST™ can provide an array of services such as on-site IQ, OQ, PQ, Customized Protocol Generation, Equipment Calibration, Asset Management, and Certification Services (US only). Please call 888-793-2300 or email VWRCATALYST™@vwr.com for further details.
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty