Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Barnstead™ MicroPure Water Purification Systems, Thermo Scientific

Supplier: Thermo Scientific

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Barnstead™ MicroPure™
50132370 50132373 50132370 50132374 50132372
89230-430EA 8800 USD InStock
89230-430 89230-434 89230-432 89230-428 CA89230-428
Barnstead™ MicroPure Water Purification Systems, Thermo Scientific
Water Purification Systems Pure and Ultra Pure Water Systems
The Barnstead MicroPure water purification systems provides up to 15L/day of Type 1, ultrapure (18.2 megohm) water from pre-treated feed water. All MicroPure systems include:feed water monitoring to protect your consumables, quick discconnects to make changing consumables easy, and all core consumables and tubing. MicroPure-ST systems come standard with a 6 liter integrated feed tank for labs that do not have a ready supply of pre-treated feed water.

The MicroPure systems offer highly versatile mounting options.The units can be mounted on the wall, on the bench or under the bench.

The MicroPure systems are offered in a variety of configurations to meet a wide range of applications needs.
The MicroPure systems are ideal for atomic absorption, ion chromatography or other analytical procedures.
The MicroPure UV systems are ideal for inorganic and organic trace analysis, HPLC, or ICP-MS.
The MicroPure UV/UF systems are ideal for molecular biology, PCR, DNA work, cell or tissue culture media.

Certifications: CSA, CE

Ordering information: Units come standard with universal power supply (90-240V, 50.60Hz), all necessary tubing to connect to water inlet supply.MicroPure-ST systems come with integrated 6L revservoir. Feed water must be pre-treated source such as deionized, distilled or treated by reverse osmosis.Alternative units are available that can make Type 1 water from tap water if a source of pre-treated feedwater is not available.

Order Now
 

ORDER

SPECIFICATIONS

ACCESSORIES

SERVICES


Services: Thermo Scientific provides several service options for Thermo Barnstead Water System such as Installation, Extended Warranty, Extended Warranty plus on-site Annual Preventative Maintenance, and Total Care. To learn more about these services please search "Thermo Barnstead Water System Technical Services" in the website search field.
In addition, VWRCATALYST™ can provide an array of services such as on-site IQ, OQ, PQ, Customized Protocol Generation, Equipment Calibration, Asset Management, and Certification Services (US only). Please call 888-793-2300 or email VWRCATALYST@vwr.com for further details.

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty