Would you like to log in again?
SP Bel-Art 80-Well Reversible PCR and Microcentrifuge Tube Racks, Bel-Art Products, a part of SP
Supplier: Bel-Art
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
F18845-0015 F18845-0008 F18845-0001 F18845-0004 F18845-0006 F18845-0012 F18845-0005 F18845-0003 F18845-0013 F18845-0016 F18845-0007 F18845-0017 F18845-0014
89230-662CS 342.95 USD InStock
89230-662 89230-672 89230-660 89230-670 89230-676 89230-654 89230-652 89230-674 89230-648 89230-658 89230-656 89230-678 89230-680
SP Bel-Art 80-Well Reversible PCR and Microcentrifuge Tube Racks, Bel-Art Products, a part of SP
Racks Tube Racks
Perfect for benchtop use or freezer storage, this double-sided polypropylene plastic rack accommodates 0.2 ml thin-walled PCR tubes on one side while the opposite side holds 1.5 ml to 2.0 ml microcentrifuge tubes.
  • Ideal compact storage solution
  • Molded-in handles
  • Color-coded racks assist in quick visual identification
  • Can be used for freezer storage
  • Autoclavable
80 wells on each side keep tubes upright in a 5 x 16 array and are numbered for easy sample identification. Twelve rack colors assist in sample organization and quick visual identification. Molded-in handles provide a secure grip for transport and a matte side panel accepts tape or direct labeling. Racks can be used for freezer storage down to –80°C (-112°F) and can be autoclaved at 121°C (250°F). Overall dimensions: 22.7 x 6.7 x 2.7 cm (815/16 x 25/8 x 11/16"). 5 per pack.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty