Would you like to log in again?
SP Bel-Art Double-Sided, 96-Well - 0.5, 1.5 and 2.0 ml MicrocentrifugeTube Racks, Bel-Art®, Bel-Art Products, a part of SP
Supplier: Bel-Art
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
F18845-0011 F18845-0009 F18845-0100 F18845-0023 F18845-0022 F18845-0020 F18845-0030 F18845-0026 F18845-0025 F18845-0021 F18845-0031 F18845-0029 F18845-0027 F18845-0019 F18845-0010 F18845-0028 F18845-0018
89230-684CS 546.22 USD InStock
89230-684 89230-694 89230-682 89230-692 89230-688 89230-666 89230-698 89230-664 89230-686 89230-696 89230-702 89230-668 89230-701 89230-706 89230-704 89230-708 89230-690
SP Bel-Art Double-Sided, 96-Well - 0.5, 1.5 and 2.0 ml MicrocentrifugeTube Racks, Bel-Art®, Bel-Art Products, a part of SP
Racks Tube Racks
Double-sided, compact polypropylene plastic racks are perfect for use on the benchtop or in the freezer.
  • 14 colors for easy organization and quick visual identification
  • Polypropylene rack construction
  • Double-sided design
  • Optional cover is translucent for easy viewing
  • Molded-in handles
One side accommodates 0.5 mL microcentrifuge tubes while the opposite side holds 1.5 mL to 2.0 mL microcentrifuge tubes. 96 wells on each side keep tubes upright in an 8 x 12 array. Alphanumeric index permits easy tube identification and a matte side panel accepts tape or direct labeling. Molded-in handles provide a secure grip for transport. Racks can be used for freezer storage down to -80°C (-112°F) and can be autoclaved at 121°C (250°F). Overall dimensions: 21.3 x 11.4 x 2.7 cmH (8-3/8 x 4-1/2 x 1-1/16"). Optional cover fits either side, protects samples and is translucent for easy viewing. Cover overall dimensions: 20.7 x 11.8 x 2.9 cmH (8-1/8 x 4-5/8 x 1-1/8"). Racks and cover: 5 per pack. Optional Cover protects samples and is translucent for viewing (89230-708).
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty