Would you like to log in again?
SP Bel-Art 96-Well PCR Tube Rack with Cover, Bel-Art Products, a part of SP
Supplier: Bel-Art
F18902-0006 F18902-0002 F18902-0005 F18902-0001 F18902-0004 F18902-0003
89230-710CS 549.77 USD InStock
89230-710 89230-720 89230-714 89230-712 89230-718 89230-716
SP Bel-Art 96-Well PCR Tube Rack with Cover, Bel-Art Products, a part of SP
Racks Tube Racks
Designed for preparing and storing 0.2 mL thin-walled PCR tubes and strip tubes before or after PCR sample processing applications.
  • Compact benchtop footprint
  • Alphanumeric grid provides easy sample identification
  • Color-coded racks allow quick visual identification
  • Translucent polypropylene cover
The compact rack features 96 wells that keep tubes securely upright in an 8 x 12 array
Each rack includes translucent polypropylene cover to protect samples and allow viewing without removal. Steam autoclavable at 121°C (250°F).
An alphanumeric grid assists in easy sample identification. Five rack colors assist in sample organization and quick visual identification.
Ordering information: The Assortment pack includes one of each color; flourescent blue, green, pink, yellow, and orange.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty