Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
SpectraDrop™ Plate Systems, Molecular Devices
Supplier: Molecular Devices
SMAX DROP 64C SMAX DROP 24 SMAX DROP 24C
89230-750EA 28394.18 USD InStock
89230-750 89230-746 89230-748
SpectraDrop™ Plate Systems, Molecular Devices
Microplate Readers
Molecular Devices’ unique SpectraDrop™ Micro-Volume Plate offers a high throughput and accurate solution for low volume measurement.
  • High throughput
  • Innovative design
  • Accelerated sample prep
  • Increased productivity
Innovative and flexible design features enable accelerated sample preparation time and increased laboratory productivity of DNA, RNA, and protein samples as low as 2 μL
The SpectraDrop™ Micro-Volume Plate incorporates a specially designed adapter and slide pair providing uniform multi-sample assembly. The two-slide design makes the cleaning process a easy and reliable. The users can quickly wipe each slide, autoclave them, sonicate or simply replace them without having to worry about any calibration steps. The simplicity of this arrangement also allows each user to have their own slide combination, making it ideal for multi-user laboratories.
The SpectraDrop™ Micro-Volume Plate comes in two density configurations, 24-wells per plate and 64-wells per plate, and two volume options, 2 or 4 μL, allowing users to select the configuration ideal for their sensitivity, volume and throughput needs. The SpectraDrop™ design never requires any calibration.
The SpectraDrop™ Micro-Volume Plate assures uniform and reproducible analysis and integrates seamlessly with all UV capable SpectraMax Readers - giving you the cost effective option of adding low volume detection with the high standard of data performance of Molecular Devices' readers.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty