Would you like to log in again?
Capto™ Core 700 Multimodal Chromatography Media
Supplier: Cytiva
Capto™
17548102 17548102 17548101 17548101
89230-830EA 1009.06 USD InStock
89230-830 CA89230-828 CA89230-830 89230-828
Capto™ Core 700 Multimodal Chromatography Media
Protein Purification Protein Purification Resins
Capto™ core 700 is designed for intermediate purification and polishing of viruses and other large biomolecules (molecular weight >700000).
  • Novel core bead technology allows efficient capture of contaminants while target molecules are collected in the flowthrough
  • Significantly improved productivity and higher flow rates compared with gel filtration methods
  • Straightforward optimization due to flowthrough chromatography and robust performance
  • Convenient small-scale purification, process development, and scale-up using prepacked HiTrap and HiScreen columns
The novel core bead technology and multimodal, octylamine ligand give dual functionality, size exclusion, and binding chromatography in one chromatography medium (resin). Available in various pack sizes for packing columns and in convenient, prepacked formats.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty