Would you like to log in again?
HiTrap Columns, Capto Core 700, Cytiva
Supplier: Cytiva
HiTrap™
17-5481-51 17548151
89230-834PK 521.24 USD InStock
CA89230-834 89230-834
HiTrap Columns, Capto Core 700, Cytiva
Chromatography Columns
HiTrap™ Capto Core 700 is prepacked with Capto Core 700, a multimodal BioProcess medium for intermediate purification and polishing of viruses and other large biomolecules.
  • High capacity and productivity in flowthrough mode
  • Wide operational window of pH and conductivity
  • Prepacked HiTrap™ columns for easy screening and convenient process development, as well as small scale purification
Capto Core 700 is a novel, layered bead chromatography resin consisting of a porous outer layer and ligand-activated core. The octylamine ligands found in the core of the beads are multimodal, being both hydrophobic and positively charged. These internalised ligands bind impurities strongly over a wide range of pH and salt concentration. The pores of the bead outer layer prevent large targets (Mr >700000) from binding to the ligands while smaller impurities can enter freely into the beads where they are captured.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty