Would you like to log in again?
Sodium Periodate Powder Pillows for Manganese, 25 ml, Hach
Supplier: Hach
98499 98499
89230-904PK 86.31 USD InStock
CA11016-478 89230-904
Sodium Periodate Powder Pillows for Manganese, 25 ml, Hach
Environmental Test Kits
For determination of manganese by the periodate oxidation method
EPA approved for reporting wastewater analyses (digestion required). Hach method 8034. Sample size: 25 mL.
Ordering information: Pack of 100 Powder Pillows. Hazardous Items: Items with this mark may be considered hazardous under some shipping conditions. If necessary, we will change your selected shipping method to accommodate these items.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty