Would you like to log in again?
Chlorine Reagent Set (Total), Ultra Low Range DPD, Hach
Supplier: Hach
2563000 2563000
89231-132EA 137.43 USD InStock
89231-132 CA11024-852
Chlorine Reagent Set (Total), Ultra Low Range DPD, Hach
Environmental Test Kits
For determination of ultralow Total Chlorine concentrations by the DPD Method
*This product has not been evaluated to test for chlorine and chloramines in medical applications in the United States.
Hach Method 8370. USEPA accepted for drinking water and wastewater analysis. Range: 2 - 500 μg/L Cl2. Reagents are ampuled for stability.
Ordering information: Reagent set includes ULR Chlorine Buffer Solution (1.5-mL ampules), DPD Indicator Solution for ULR Chlorine (1.5-mL ampules) and Blanking Reagent for 20 tests.
Hazardous Items:
Items with this mark may be considered hazardous under some shipping conditions.
If necessary, we will change your selected shipping method to accomodate these items.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty