Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
IntelliCAL™ LDO101 Standard Luminescent Dissolved Oxygen (LDO) Probe, 3 meter cable, Hach
Supplier: Hach
LDO10103
89231-212EA 2029.46 USD InStock
89231-212
IntelliCAL™ LDO101 Standard Luminescent Dissolved Oxygen (LDO) Probe, 3 meter cable, Hach
Electrodes Dissolved Oxygen Electrodes
IntelliCAL™ LDO101 is a digital, luminescent dissolved oxygen (LDO) probe.
  • Hach IntelliCAL™ LDO is a revolutionary breakthrough in the measurement of dissolved oxygen, eliminating the numerous reliability and maintenance concerns inherent of older DO measurement technologies.
  • IntelliCAL™ digital probes provide ultimate traceability in measurement history.
  • IntelliCAL™ digital probes alert the user when recalibration is needed.
  • IntelliCAL™ digital probes can be moved between meters without the need to recalibrate or reenter measurement settings.
The standard version is available with a 3 or 1 meter cable and is intended for laboratory use. The LDO101 is ideal for measuring dissolved oxygen in wastewater, drinking water and general water quality applications.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty