Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
IntelliCAL™ LDO101 Rugged Luminescent/Optical Dissolved Oxygen (LDO) Probe, Hach
Supplier: Hach
LDO10130 LDO10130 LDO10115 LDO10115
89231-214EA 3239.2 USD InStock
CA11016-320 89231-214 89231-216 CA11014-086
IntelliCAL™ LDO101 Rugged Luminescent/Optical Dissolved Oxygen (LDO) Probe, Hach
Electrodes Dissolved Oxygen Electrodes
Hach IntelliCAL™ LDO offers a revolutionary breakthrough in the measurement of dissolved oxygen, eliminating the numerous reliability and maintenance concerns inherent of older DO measurement technologies
IntelliCAL™ LDO101 is a digital, luminescent/optical dissolved oxygen (LDO) probe. The rugged version is available with a 5, 10, 15, or 30 meter cable and is intended for field use. The LDO101 is ideal for measuring dissolved oxygen in wastewater, drinking water and general water quality applications.
IntelliCAL™ Rugged digital probes can reliably test in the harshest environmental conditions. IntelliCAL™ digital probes provide ultimate traceability in measurement history. IntelliCAL™ digital probes alert the user when re-calibration is needed. IntelliCAL™ digital probes can be moved between meters without the need to re-calibrate or re-enter measurement settings.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty