MicroLiter µLPlate® components include a proprietary product line of 96-well round and square well microplates with glass inserts and color-coded caps.
- Variety of volumes available
- Standard chromatography closures
- Caps with septa
- Silicone mats are resealable
A comprehensive line of 96-Well microplates for chromatography, the Plate Sampling System™ uses closures familiar to those who work with standard chromatography vials
Polypropylene μLPlates® have a variety of closures that will stabilize the sample during the analytical sample run and, if necessary, reseal after the sample has been injected. Users may also choose the security of a stopper coupled with the strength of a mechanical closure system. The mechanical closure system allows even GC samples to be converted to 96-Well format, by sealing the limited volume insert on the inside surface of the insert and compressing the PTFE/Silicone rubber using a mechanical seal. A variety of insert options allows users to choose the specifications appropriate for any application.
The system combines microplates, glass inserts, μLmats™, and caps with chromatography-based septa to offer secure vessels to maintain even the most volatile samples.