Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

SilTite® μ-Unions, Gas Chromatography Connections, Trajan Scientific and Medical

Supplier: Trajan Scientific and Medical
SilTite™
073567 073562 073560 073563 073568 073561 073569 073566 073565 073571 073574 073564 073570 073572 073573
89232-108EA 311.3 USD InStock
89232-108 89232-118 89232-106 89232-128 89232-116 89232-126 89232-114 89232-124 89232-112 89232-134 89232-122 89232-110 89232-132 89232-120 89232-130
SilTite® μ-Unions, Gas Chromatography Connections, Trajan Scientific and Medical
Fittings and Connectors
Your chromatography analysis does not end with the selection of the GC column

  • Inert
  • Zero dead volume
  • Optimized peak shapes
  • Robust and stable
  • For both extreme temperature and pressure cycling
  • Easy to install
  • Leak-free
  • No need to re-tighten after installation
  • Only 9 mm in total length
  • Mass of \<0.5 g
  • Low thermal mass

These micro unions feature chemically deactivated stainless steel ferrules alleviating active sites. They should be used when fitting a pre-column or guard column, GC to GC connections.

The combination of components selected for your instrument also make an important contribution to successful separations. SGE brings technology and chromatography expertise to improve your chromatography. SGE manufactures an array of connection types for GC use. Poorly defined or selected connections can lead to an increase in dead volume, leaks from cycled temperatures and mismatched tubing sizes. SGE SilTite™ μ-Unions are specifically designed and precision engineered for use in capillary GC.

Certifications: RoHS compliant per Directive 2011/65/EU

Ordering information: SilTite ?-Unions and replacement ferrules are available in sizes to connect 0.1 - 0.25 mm ID, 0.32 mm I.D. and 0.53 mm I.D. columns and fused silica tubing. Packs contain 2 unions, 5 double taper ferrules and the necessary tools.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty