Would you like to log in again?
SilTite™ FingerTite GC Ferrule Systems, Trajan Scientific and Medical
Supplier: Trajan Scientific and Medical
SilTite™ FingerTite
073611 073331 0736101 073228 073629 073230 073628 073229 073619 073630 073618 073633 073219 07361054 073473 0736105 07362053 073554 073622 073612 073615 0736104 073620 073330 073616 073623 073636 07361053 073640 073632 073332 1034085 0736103 073621 073614 073631 0736102 073625 0736100 073610
10799-824PK 124.78 USD InStock
10799-824 89232-150 89232-170 10799-820 89232-190 10799-822 14226-536 97051-960 97051-962 10799-794 89232-136 97051-964 10799-796 89232-178 97051-966 89232-154 89232-176 97051-946 89232-152 89232-196 97051-948 89232-182 10799-812 89232-160 10799-814 10799-810 10799-816 97051-950 97051-952 89232-148 97051-954 89232-146 89232-168 97051-956 89232-166 89232-188 89232-144 97051-958 89232-142 89232-140
SilTite™ FingerTite GC Ferrule Systems, Trajan Scientific and Medical
Fittings and Connectors
SilTite FingerTite is a GC ferrule system that has been designed to be installed only with the force of you fingers, delivering an easy, leak and maintenance free seal.
  • Simplified installation process
  • Leak free
  • No column creep
  • Cannot overtighten
  • No broken columns
  • Chemically inert
  • No contamination
  • Metal ferrules
  • No temperature limit
  • No tools required
These ferrules are for use with MS, high throughput laboratories. Starter kits are available for Agilent, PerkinElmer, Shimadzu, Thermo Scientific and Varian/Bruker MC-MS systems.
Certifications: RoHS compliant per Directive 2011/65/EU.
Ordering information: Each starter kit includes all the parts necessary to convert one GC system (one injector and one detector) to the SilTite FingerTite system. In addition there are 5 SilTite FingerTite nuts, one packet (10 ferrules) of 0.4 mm ID SilTite FingerTite ferrules (suitable for installing 0.1 - 0.25 mm ID capillary columns) and a ferrule install tool which allows you to seat the ferrule in the correct position on the capillary column.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty