Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

VWR® Wooden Inoculation Picks

Supplier: VWR
VWR-89233-132 VWR-89233-136 VWR-89233-130 VWR-89233-140 VWR-89233-134 VWR-89233-138
89233-136CS 1989.47 USD InStock
89233-136 89233-138 89233-132 89233-134 89233-140 89233-130
VWR® Wooden Inoculation Picks
Spreaders Inoculating Loops and Needles
VWR® wooden inoculation picks are available in three convenient sizes (1.28”, 2.5”, and 6”) to accommodate various transfer, inoculating, and streaking needs.

  • Autoclavable
  • 100% USA grown and manufactured
  • Environmentally friendly
  • Naturally biodegradable

VWR® mini picks are 1.28" long and are sized to fit in a closed 1.5 mL tube

VWR® standard picks are 2.5" long single ended toothpicks. VWR® large picks are 6" long natural wood alternatives to plastic inoculation picks.

Mini picks eliminate wasted time and effort. Simply pick, drop, and close.

Certifications: Meet FDA food contact standards. ISO 9001:2008, ISO 13485:2003 and FDA registered facility.

Ordering information: Picks are available with semi (SP) or needlepoint (NP) tips.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty